Learn More
Biomatik Corporation IL-6 (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51904
Encompass_Preferred
IL-6 (Murine) Sandwich ELISA Kit for detecting IL-6 in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 62-4000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P08505. Gene ID: 16193. Target Synonyms: Interleukin-6; B-cell hybridoma growth factor; Interleukin HP-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9130