Learn More
Biomatik Corporation E-Cadherin (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51933
Encompass_Preferred
E-Cadherin (Human) Sandwich ELISA Kit for detecting E-Cadherin in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 188-12000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P12830. Gene ID: 999. Target Synonyms: CAM 120/80; Epithelial cadherin; Uvomorulin; CD324; CDHE; UVO. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9142