Learn More
Biomatik Corporation IL-19 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51957
Encompass_Preferred
IL-19 (Human) Sandwich ELISA Kit for detecting IL-19 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9UHD0. Gene ID: 29949. Target Synonyms: Interleukin-19; Melanoma differentiation-associated protein-like protein; NG.1; ZMDA1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9148