Learn More
Biomatik Corporation Pentraxin-3 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52057
Encompass_Preferred
Pentraxin-3 (Human) Sandwich ELISA Kit for detecting Pentraxin-3 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 219-14000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P26022. Gene ID: 5806. Target Synonyms: Pentraxin-related protein PTX3; Pentaxin-related protein PTX3; Tumor necrosis factor alpha-induced protein 5; TNF alpha-induced protein 5; Tumor necrosis factor-inducible gene 14 protein; TSG-14; TNFAIP5; TSG14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9175