Learn More
Biomatik Corporation Human 2, 5-OAS (2, 5-oligoadenylate synthetase) ELISA Kit
Supplier:  Biomatik Corporation EKF58287
Encompass_Preferred
Human 2, 5-OAS (2, 5-oligoadenylate synthetase) ELISA Kit for detecting 2, 5-OAS in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.469-30ng/ml. Sensitivity: 0.281ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P00973. Target Synonyms: 2'-5'-oligoadenylate synthase 1; (2-5')oligo(A) synthase 1; 2-5A synthase 1; E18/E16; p46/p42 OAS; OIAS; OAS1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8088