Learn More
Biomatik Corporation Rat SNAP-25 (Synaptosome Associated Protein 25) ELISA Kit
Supplier:  Biomatik Corporation EKF58391
Encompass_Preferred
Rat SNAP-25 (Synaptosome Associated Protein 25) ELISA Kit for detecting SNAP-25 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P60881. Target Synonyms: SNAP25; Synaptosomal-associated protein 25; SNAP-25; Super protein; SUP; Synaptosomal-associated 25 kDa protein; SNAP; RIC4; SEC9; SNAP; RIC-4; SNAP-25. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8111