Learn More
Biomatik Corporation B-RAF Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50067
Encompass_Preferred
B-RAF Colorimetric Cell-Based ELISA Kit for detecting B-RAF in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P15056. Gene ID: 673. Target Synonyms: B-RAF; B-RAF proto-oncogene serine/threonine-protein kinase; B-Raf proto-oncogene serine/threonine-protein kinase; BRAF; BRAF1; c-RMIL; kinase B-Raf; p94; RAFB1; RMIL; RMIL serine/threonine-protein kinase; v-Raf murine sarcoma viral oncogene homolog B1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8902