Learn More
Biomatik Corporation Mouse Cross Linked C-Telopeptide Of Type I Collagen (CTXI) ELISA Kit
Supplier:  Biomatik Corporation EKL54194
Encompass_Preferred
Mouse Cross Linked C-Telopeptide Of Type I Collagen (CTXI) ELISA Kit for detecting CTXI in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.046ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Target Synonyms: CTX-I; CTX1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8584