Learn More
Biomatik Corporation Rat Fatty Acid Binding Protein 3, Muscle And Heart (FABP3) ELISA Kit
Supplier:  Biomatik Corporation EKL54242
Encompass_Preferred
Rat Fatty Acid Binding Protein 3, Muscle And Heart (FABP3) ELISA Kit for detecting FABP3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P07483. Gene ID: 79131. Target Synonyms: MDGI; FABP11; H-FABP; O-FABP; Hfabp; Mammary Derived Growth Inhibitor; Fatty Acid Binding Protein 11; Muscle fatty acid-binding protein; Heart-type fatty acid-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8589