Learn More
Biomatik Corporation BD-4 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51821
Encompass_Preferred
BD-4 (Human) Sandwich ELISA Kit for detecting BD-4 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 8-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8WTQ1. Gene ID: 140596; 503618. Target Synonyms: HBD-4; DEFB-4; HBD4; DEFB104B; Beta-defensin 4; BD-4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9105