Learn More
Biomatik Corporation EGF (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51828
Encompass_Preferred
EGF (Human) Sandwich ELISA Kit for detecting EGF in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 8-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P01133. Gene ID: 1950. Target Synonyms: Epidermal growth factor; Urogastrone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9107