Learn More
Biomatik Corporation IL-10 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51840
Encompass_Preferred
IL-10 (Human) Sandwich ELISA Kit for detecting IL-10 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P22301. Gene ID: 3586. Target Synonyms: Interleukin-10; Cytokine synthesis inhibitory factor; CSIF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9114