Learn More
Biomatik Corporation IL-8 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51859
Encompass_Preferred
IL-8 (Human) Sandwich ELISA Kit for detecting IL-8 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P10145. Gene ID: 3576. Target Synonyms: Interleukin-8; C-X-C motif chemokine 8; Emoctakin; Granulocyte chemotactic protein 1; GCP-1; Monocyte-derived neutrophil chemotactic factor; MDNCF; Monocyte-derived neutrophil-activating peptide; MONAP; Neutrophil-activating protein 1; NAP-1; Protein 3-10C; T-cell chemotactic factor; CXCL8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9119