Learn More
Biomatik Corporation IL-1alpha (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51898
Encompass_Preferred
IL-1alpha (Murine) Sandwich ELISA Kit for detecting IL-1alpha in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 16-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P01582. Gene ID: 16175. Target Synonyms: Interleukin-1 alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9128