Learn More
Biomatik Corporation TGFbeta-R3 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51982
Encompass_Preferred
TGFbeta-R3 (Human) Sandwich ELISA Kit for detecting TGFbeta-R3 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 157-10000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q03167. Gene ID: 7049. Target Synonyms: Transforming growth factor beta receptor type 3; Betaglycan; Transforming growth factor beta receptor III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9159