Learn More
Biomatik Corporation ACE (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52007
Encompass_Preferred
ACE (Human) Sandwich ELISA Kit for detecting ACE in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P12821. Gene ID: 1636. Target Synonyms: Angiotensin-converting enzyme; Dipeptidyl carboxypeptidase I; Kininase II; CD143; DCP; DCP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9163