Learn More
Biomatik Corporation BD-1 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51818
Encompass_Preferred
BD-1 (Human) Sandwich ELISA Kit for detecting BD-1 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 4-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P60022. Gene ID: 1672. Target Synonyms: Beta-defensin 1; BD-1; Defensin beta 1; hBD-1; HBD1; HBP1; DEFB1; HBD-1; HBP-1; DEFB101; DEFB-1; MGC51822. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9102