Learn More
Biomatik Corporation Canine acetylcholine receptor antibody, AChRab ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC31831
Encompass_Preferred
The Canine acetylcholine receptor antibody, AChRab ELISA Kit allows for quantitative determination of acetylcholine receptor antibody, AChRab in serum, plasma. Species Reactivity: Dog. Detection Range: 3.75 ng/mL-240 ng/mL. Sensitivity: 0.94 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-7522