Learn More
Biomatik Corporation PAI-1 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51871
Encompass_Preferred
PAI-1 (Human) Sandwich ELISA Kit for detecting PAI-1 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 23-3000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P05121. Gene ID: 5054. Target Synonyms: Plasminogen activator inhibitor 1; Endothelial plasminogen activator inhibitor; Serpin E1; PLANH1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9123