Learn More
Biomatik Corporation TGF-beta3 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51980
Encompass_Preferred
TGF-beta3 (Human) Sandwich ELISA Kit for detecting TGF-beta3 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P10600. Gene ID: 7043. Target Synonyms: Transforming growth factor beta-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9157