Learn More
Biomatik Corporation Granzyme B (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51994
Encompass_Preferred
Granzyme B (Murine) Sandwich ELISA Kit for detecting Granzyme B in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 40-5000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P04187. Gene ID: 14939. Target Synonyms: Granzyme B(G,H); CTLA-1; Cytotoxic cell protease 1; Fragmentin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9162