Learn More
Biomatik Corporation CRP (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52030
Encompass_Preferred
CRP (Human) Sandwich ELISA Kit for detecting CRP in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P02741. Gene ID: 1401. Target Synonyms: CRP; PTX1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9167