Learn More
Biomatik Corporation Ku70/XRCC6 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50348
Encompass_Preferred
Ku70/XRCC6 Colorimetric Cell-Based ELISA Kit for detecting Ku70/XRCC6 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P12956. Gene ID: 2547. Target Synonyms: 70 kDa subunit of Ku antigen ATP-dependent DNA helicase II 70 kDa subunit; CTC box binding factor 75 kDa subunit; CTC75; CTCBF; G22P1; Lupus Ku autoantigen protein p70; TLAA; Thyroid-lupus autoantigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0619