Learn More
Biomatik Corporation MAGE-1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50356
Encompass_Preferred
MAGE-1 Colorimetric Cell-Based ELISA Kit for detecting MAGE-1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P43355; Q8WWH6. Gene ID: 4100. Target Synonyms: MAGE1; MAGE1A; MZ2-E; melanoma-associated antigen 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0626