Learn More
Biomatik Corporation Canine Cardiac Troponin Ⅰ (cTn-Ⅰ) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC31840
Encompass_Preferred
The Canine Cardiac Troponin Ⅰ (cTn-Ⅰ) ELISA Kit allows for quantitative determination of Cardiac Troponin Ⅰ (cTn-Ⅰ) in serum, plasma, tissue homogenates. Species Reactivity: Dog. Detection Range: 23.5 pg/mL-1500 pg/mL. Sensitivity: 5.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-7524