Learn More
Biomatik Corporation MAPKAPK2 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50363
Encompass_Preferred
MAPKAPK2 Colorimetric Cell-Based ELISA Kit for detecting MAPKAPK2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P49137. Gene ID: 9261. Target Synonyms: MAP kinase-activated protein kinase 2; MAPK-activated protein kinase 2; MAPK2; MAPKAP kinase 2; MAPKAPK-2; MAPKAPK2; RPS6KC1; kinase MAPKAPK2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0632