Learn More
Biomatik Corporation MMP-1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50389
Encompass_Preferred
MMP-1 Colorimetric Cell-Based ELISA Kit for detecting MMP-1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P03956. Gene ID: 4312. Target Synonyms: Interstitial collagenase; Fibroblast collagenase; Matrix metalloproteinase-1; MMP-1; MMP1; CLG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0656