Learn More
Biomatik Corporation MSK2 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50406
Encompass_Preferred
MSK2 Colorimetric Cell-Based ELISA Kit for detecting MSK2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O75676. Gene ID: 8986. Target Synonyms: Nuclear mitogen- and stress-activated protein kinase 2; 90 kDa ribosomal protein S6 kinase 4; Ribosomal protein kinase B; RSKB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0670