Learn More
Biomatik Corporation Pax-5 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50461
Encompass_Preferred
Pax-5 Colorimetric Cell-Based ELISA Kit for detecting Pax-5 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q02548. Gene ID: 5079. Target Synonyms: Paired box protein Pax-5; B-cell-specific transcription factor; BSAP; PAX5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0715