Learn More
Biomatik Corporation PLCG1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50493
Encompass_Preferred
PLCG1 Colorimetric Cell-Based ELISA Kit for detecting PLCG1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P19174. Gene ID: 5335. Target Synonyms: 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-1; PLC-148; Phosphoinositide phospholipase C-gamma-1; Phospholipase C-II; PLC-II; Phospholipase C-gamma-1; PLC-gamma-1; PLCG1; PLC1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0745