Learn More
Biomatik Corporation Retinoid X Receptor γ Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50523
Encompass_Preferred
Retinoid X Receptor γ Colorimetric Cell-Based ELISA Kit for detecting Retinoid X Receptor γ in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P48443. Gene ID: 6258. Target Synonyms: Retinoic acid receptor RXR-gamma; Nuclear receptor subfamily 2 group B member 3; Retinoid X receptor gamma; RXRG; NR2B3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0768