Learn More
Biomatik Corporation SENP1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50532
Encompass_Preferred
SENP1 Colorimetric Cell-Based ELISA Kit for detecting SENP1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9P0U3. Gene ID: 29843. Target Synonyms: Sentrin-specific protease 1; Sentrin/SUMO-specific protease SENP1; SENP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0777