Learn More
Biomatik Corporation SF1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50538
Encompass_Preferred
SF1 Colorimetric Cell-Based ELISA Kit for detecting SF1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15637. Gene ID: 7536. Target Synonyms: BBP; CW17; mammalian branch point binding protein mBBP; mZFM; SF01; Splicing factor 1; splicing factor SF1; transcription factor ZFM1; ZFM1; ZFP162; zinc finger gene in MEN1 locus; zinc finger protein 162; ZNF162. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0783