Learn More
Biomatik Corporation Survivin Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50573
Encompass_Preferred
Survivin Colorimetric Cell-Based ELISA Kit for detecting Survivin in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O15392. Gene ID: 332. Target Synonyms: API4; apoptosis inhibitor 4; apoptosis inhibitor survivin; baculoviral IAP repeat-containing protein 5; BIRC5; IAP4; TIAP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0810