Learn More
Biomatik Corporation TIE2 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50587
Encompass_Preferred
TIE2 Colorimetric Cell-Based ELISA Kit for detecting TIE2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q02763. Gene ID: 7010. Target Synonyms: Tyrosine-protein kinase receptor TIE-2; hTIE2; Tyrosine-protein kinase receptor TEK; p140 TEK; Tunica interna endothelial cell kinase; CD202b antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0822