Learn More
Biomatik Corporation TRADD Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50599
Encompass_Preferred
TRADD Colorimetric Cell-Based ELISA Kit for detecting TRADD in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15628. Gene ID: 8717. Target Synonyms: Tumor necrosis factor receptor type 1-associated DEATH domain protein; TNFR1-associated DEATH domain protein; TNFRSF1A-associated via death domain; TRADD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0833