Learn More
Biomatik Corporation Tubulin α Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50606
Encompass_Preferred
Tubulin α Colorimetric Cell-Based ELISA Kit for detecting Tubulin α in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q71U36; P68363. Gene ID: 7846; 10376; 84790; 113457; 7278; 112714; 7277. Target Synonyms: Alpha-tubulin 1; TBA1; TBA1A; TBA4A; Testis-specific alpha- tubulin; TUBA1; Tubulin alpha-1 chain; Tubulin H2-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0840