Learn More
Biomatik Corporation VAV2 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50616
Encompass_Preferred
VAV2 Colorimetric Cell-Based ELISA Kit for detecting VAV2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P52735. Gene ID: 7410. Target Synonyms: Vav-2 protein; Guanine nucleotide exchange factor VAV2; VAV-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0848