Learn More
Biomatik Corporation VCP Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50618
Encompass_Preferred
VCP Colorimetric Cell-Based ELISA Kit for detecting VCP in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P55072. Gene ID: 7415. Target Synonyms: 15S Mg(2+)-ATPase p97 subunit; TER ATPase; TERA; Transitional endoplasmic reticulum ATPase; Valosin containing protein; VCP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0850