Learn More
Biomatik Corporation XPF Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50630
Encompass_Preferred
XPF Colorimetric Cell-Based ELISA Kit for detecting XPF in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q92889. Gene ID: 2072. Target Synonyms: DNA excision repair protein ERCC-4; DNA repair endonuclease XPF; excision repair cross-complementing rodent repair deficiency complementation group 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0859