Learn More
Biomatik Corporation MAST3 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50664
Encompass_Preferred
MAST3 Colorimetric Cell-Based ELISA for detecting MAST3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O60307. Gene ID: 23031. Target Synonyms: Microtubule-associated serine/threonine-protein kinase 3; MAST3; KIAA0561. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0890