Learn More
Biomatik Corporation DLX3 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50666
Encompass_Preferred
DLX3 Colorimetric Cell-Based ELISA for detecting DLX3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O60479. Gene ID: 1747. Target Synonyms: Homeobox protein DLX-3; DLX3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0892