Learn More
Biomatik Corporation Musculin Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50671
Encompass_Preferred
Musculin Colorimetric Cell-Based ELISA for detecting Musculin in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O60682. Gene ID: 9242. Target Synonyms: Musculin; Activated B-cell factor 1; ABF-1; MSC; ABF1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0897