Learn More
Biomatik Corporation PFDN1 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50673
Encompass_Preferred
PFDN1 Colorimetric Cell-Based ELISA for detecting PFDN1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O60925. Gene ID: 5201. Target Synonyms: Prefoldin subunit 1; PFD1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0899