Learn More
Biomatik Corporation TRIM3 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50675
Encompass_Preferred
TRIM3 Colorimetric Cell-Based ELISA for detecting TRIM3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O75382. Gene ID: 10612. Target Synonyms: Tripartite motif-containing protein 3; RING finger protein 22; Brain-expressed RING finger protein; RING finger protein 97; TRIM3; BERP; RNF22; RNF97. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0901