Learn More
Biomatik Corporation KAPCB Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50720
Encompass_Preferred
KAPCB Colorimetric Cell-Based ELISA for detecting KAPCB in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P22694. Gene ID: 5567. Target Synonyms: KAPB; KAPCB; PKA C-beta; PKA-beta; cAMP-dependent protein kinase; beta-catalytic subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0946