Learn More
Biomatik Corporation IP3KA Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50722
Encompass_Preferred
IP3KA Colorimetric Cell-Based ELISA for detecting IP3KA in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P23677. Gene ID: 3706. Target Synonyms: Inositol-trisphosphate 3-kinase A; Inositol 1,4,5-trisphosphate 3-kinase A; IP3 3-kinase A; IP3K A; InsP 3-kinase A ; ITPKA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0948