Learn More
Biomatik Corporation MARK3 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50727
Encompass_Preferred
MARK3 Colorimetric Cell-Based ELISA for detecting MARK3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P27448. Gene ID: 4140. Target Synonyms: C-TAK1; Cdc25C- associated protein kinase 1; MAP/microtubule affinity-regulating kinase 3; Protein kinase STK10; Ser/Thr protein kinase PAR-1; Serine/threonine protein kinase p78; cTAK1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0953