Learn More
Biomatik Corporation Keratin 19 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50339
Encompass_Preferred
Keratin 19 Colorimetric Cell-Based ELISA Kit for detecting Keratin 19 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P08727. Gene ID: 3880. Target Synonyms: Keratin type I cytoskeletal 19; Cytokeratin-19; CK-19; K19; KRT19. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0610