Learn More
Biomatik Corporation Keratin 5 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50341
Encompass_Preferred
Keratin 5 Colorimetric Cell-Based ELISA Kit for detecting Keratin 5 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P13647. Gene ID: 3852. Target Synonyms: Keratin type II cytoskeletal 5; Cytokeratin-5; CK-5; K5; 58 kDa cytokeratin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0612